Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Nunc™ Storage Vials with Caps
Thermo Scientific™

Nunc™ Storage Vials with Caps

Assure safe centrifugation and storage of PCR reagents, restriction enzymes, and other diagnostic and biochemical reagents or samples by using these storage vials with caps.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호부피(미터법) 작동
2643002.0 mL
2642620.5 mL
카탈로그 번호 264300
제품 가격(KRW)
330,000
Case of 500
부피(미터법) 작동:
2.0 mL
제품 가격(KRW)
330,000
Case of 500

Assure safe centrifugation and storage of PCR reagents, restriction enzymes and other biochemical reagents and samples by using Thermo Scientific™ Nunc™ Storage Vials with Caps. Available in 0.5mL and 2.0mL sizes with frosted writing area, the vials have excellent chemical resistance with leakproof closure.

Features:

  • Made from high-purity polypropylene copolymer with excellent chemical resistance
  • Leakproof high-density polyethylene screw closure
  • Centrifuged to a maximum of 13,000 x g
  • Temperature range: -40°C to 95°C
  • RNase/DNase-free

Ordering Information:

Racks 264263 and 270972 are compatible with these vials. Color coders available: 254416, 264335, 264408, 264440, 264459, 264467, 264491.

사양
인증/적합성Certificate of Analysis (COA), Certificate of Conformance (COC)
마개 재질Polyethylene
케이스당 수500
팩당 수500
제품 유형Storage Vial with Cap
부피(미터법) 전체2.2 mL
부피(미터법) 작동2.0 mL
마개 유형Screw Cap
물질PPCO
제품라인Nunc
수량500 Cs.
멸균Sterile
부피(미터법)2.2 mL
Unit SizeCase of 500