Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Nunc™ Microplate Lids
Thermo Scientific™

Nunc™ Microplate Lids

Protect samples from contamination and evaporation during assay processing, incubation or storage with these microplate lids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlate CompatibilitySterilityCondensation RingsCut-off Corners
264616384-well PlateSterileNoLong Side
250002All MicroplatesSterileNoNo
250003All MicroplatesNon-sterileNoNo
250005All MicroplatesNon-sterileNoNo
253623All MicroplatesNon-sterileYesNo
253624All MicroplatesSterileYesNo
26333996-well PlateNon-sterileYesLong Side
26412296-well PlateSterileYesLong Side
264611384-well PlateSterileNoLong Side
264612384-well PlateNon-sterileNoLong Side
26462396-well PlateSterileYesLong Side
Catalog number 264616
Price (USD)
465.86
Online Exclusive
543.25
Save 77.39 (14%)
Case of 180
Add to cart
Plate Compatibility:
384-well Plate
Sterility:
Sterile
Condensation Rings:
No
Cut-off Corners:
Long Side
Price (USD)
465.86
Online Exclusive
543.25
Save 77.39 (14%)
Case of 180
Add to cart
Protect samples from contamination and evaporation during assay processing, incubation or storage with Thermo ScientificTM NuncTM Microplate Lids.
  • Lids are for use with any standard plates
  • 96 Well Lids have individual condensation rings
  • Lids have notches for quick orientation
  • Universal Lids can be used on any standard height plate on the market
WARNING: Cancer – www.P65Warnings.ca.gov
Specifications
Dimensions (L x W)5 x 3.35 in. (127 x 85 mm)
Dimensions (L x W x H)5 x 3.35 x 0.3 in. (127 x 85 x 7.9 mm)
Evaporation BarrierYes
Height (English)0.3 in.
Height (Metric)7.9 mm
Length (English)5 in.
Length (Metric)127 mm
No. per Case180
No. per Pack20
Product TypePlate Lid
Width (English)3.35 in.
Width (Metric)85 mm
AdhesiveNone
ColorClear
Condensation RingsNo
Cut-off CornersLong Side
MaterialPolystyrene
No. of Wells384
Plate Compatibility384-well Plate
Product LineNunc
SterilitySterile
Temperature Range (Metric)-20°C to +80°C
Well ShapeAll well shapes
Unit SizeCase of 180