When screening DNA libraries, how much agar should I add to the Nunc OmniTray Single-Well Plate?
Our Nunc OmniTray Single-Well Plate can hold 35 mL of growth media. We recommend adding 1.5% agar to the growth media.
Search
View additional product information for Nunc™ OmniTray™ Single-Well Plate - FAQs (140156, 165218, 242811, 264728)
1 product FAQs found