Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals
Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium hydrogen phosphate trihydrate, 98%, extra pure, Thermo Scientific Chemicals

CAS: 7782-75-4 | Molecular Formula: H7MgO7P | Molecular Weight: 174.33 g/mol
Catalog NumberQuantityPackaging
2647800101 kgPlastic bottle
Catalog number 264780010
Price (JPY)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS7782-75-4
IUPAC Namemagnesium(2+) trihydrate hydrogen phosphate
Molecular FormulaH7MgO7P
InChI KeyOKIWLDVQGKRUNR-UHFFFAOYSA-L
SMILESO.O.O.[Mg++].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration Complexometric>=97.5 %
Chloride (Cl)=<200 ppm
Heavy metals (as Pb)=<20 ppm
Appearance (Color)White
Appearance (Form)Powder
View more

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only