Tris-maleate, 98%
Tris-maleate, 98%
Tris-maleate, 98%
Thermo Scientific Chemicals

Tris-maleate, 98%

CAS: 72200-76-1 | C4H11NO3·C4H4O4 | 237.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 264970250
Price (USD)
42.60
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
42.60
Each
Chemical Identifiers
CAS72200-76-1
IUPAC Name(2Z)-but-2-enedioic acid; bis(2-amino-2-(hydroxymethyl)propane-1,3-diol)
Molecular FormulaC12H26N2O10
InChI KeyPOSZUTFLHGNLHX-KSBRXOFISA-N
SMILESNC(CO)(CO)CO.NC(CO)(CO)CO.OC(=O)C=C/C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point104°C to 106°C
Appearance (Color)White
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only