Thermo Scientific™

Hypersil GOLD™ PEI HILIC HPLC Columns

Catalog number: 26502-202130
Thermo Scientific™

Hypersil GOLD™ PEI HILIC HPLC Columns

Catalog number: 26502-202130
Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Thermo Scientific™ Hypersil GOLD™ PEI HILIC (hydrophilic interaction liquid chromatography) LC Columns. These columns deliver straightforward separation without the need for sample derivatization or ion-pair reagents. Featuring outstanding peak shape and selectivity, the columns provide improved sensitivity for MS detection. With 1.9, 3 and 5µm particle sizes, Hypersil GOLD PEI HILIC Columns are ideal for applications within drug discovery, food testing and environmental analysis.
 
Catalog Number
26502-202130
Unit Size
Each
Particle Size
1.9 μm
Diameter (Metric)
2.1 mm
Length (Metric)
200 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)200 mm
Max. Pressure18130 psi (1250 bar)
Particle Size1.9 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
50 mm
Price (USD)
804.30
Your Price:
Availability
***
Full specifications
Quantity
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
50 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
100 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
150 mm
Price (USD)
885.15
Your Price:
Availability
***
Full specifications
Quantity
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
150 mm
Price (USD)
842.10
Your Price:
Availability
***
Full specifications
Quantity
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
2.1 mm
Length (Metric)
250 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
-
Particle Size
5 μm
Diameter (Metric)
4 mm
Length (Metric)
250 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatAnalytical Column
Diameter (Metric)4 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit Size-
Catalog Number
Unit Size
Each
Particle Size
5 μm
Diameter (Metric)
4.6 mm
Length (Metric)
250 mm
Price (USD)
886.20
Your Price:
Availability
***
Full specifications
Quantity
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
1.9 μm
Diameter (Metric)
2.1 mm
Length (Metric)
150 mm
Price (USD)
Availability
***
Full specifications
Quantity
-
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)150 mm
Max. Pressure18130 psi (1250 bar)
Particle Size1.9 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
Showing 10 of 23Show more
Catalog NumberSpecificationsUnit SizeParticle SizeDiameter (Metric)Length (Metric)Price (USD)AvailabilityQuantity
26502-202130Full specifications
Each1.9 μm2.1 mm200 mmContact Us ›***-
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)200 mm
Max. Pressure18130 psi (1250 bar)
Particle Size1.9 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
26505-052130Full specifications
Each 5 μm2.1 mm50 mm
804.30
Your Price:
***
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
26505-054630Full specifications
-5 μm4.6 mm50 mmRequest A Quote***-
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)50 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit Size-
26505-102130Full specifications
-5 μm2.1 mm100 mmRequest A Quote***-
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)100 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit Size-
26505-152130Full specifications
Each5 μm2.1 mm150 mm
885.15
Your Price:
***
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
26505-154630Full specifications
Each 5 μm4.6 mm150 mm
842.10
Your Price:
***
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)150 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
26505-252130Full specifications
-5 μm2.1 mm250 mmRequest A Quote***-
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit Size-
26505-254030Full specifications
-5 μm4 mm250 mmRequest A Quote***-
Column FormatAnalytical Column
Diameter (Metric)4 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit Size-
26505-254630Full specifications
Each5 μm4.6 mm250 mm
886.20
Your Price:
***
Column FormatAnalytical Column
Diameter (Metric)4.6 mm
Length (Metric)250 mm
Max. Pressure5800 psi (400 bar)
Particle Size5 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
26502-152130Full specifications
Each 1.9 μm2.1 mm150 mmContact Us ›***-
Column FormatAnalytical Column
Diameter (Metric)2.1 mm
For Use WithHILIC, ESI-MS
Length (Metric)150 mm
Max. Pressure18130 psi (1250 bar)
Particle Size1.9 μm
pH2 to 8
Pore Size175 Å
Surface Area220 m²/g
Temperature60°C
Stationary PhasePolyethyleneimine
Product LineHypersil GOLD
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Carbon Load6%
Column TypeHILIC
EndcappedNo
PhaseHILIC
TypeAnalytical Column
Unit SizeEach
Showing 10 of 23Show more
About the Hypersil GOLD Family of HPLC Columns:

Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5µm particle sizes for all your separation needs
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Figures