Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, Thermo Scientific Chemicals
Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, Thermo Scientific Chemicals
Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl trans-2-amino-1-cyclohexanecarboxylate hydrochloride, 98%, Thermo Scientific Chemicals

CAS: 28250-14-8 | Molecular Formula: C9H17NO2·HCl | Molecular Weight: 207.7 g/mol
Have Questions?
Catalog NumberQuantity
2656500101 g
Catalog number 265650010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS28250-14-8
IUPAC Name(1R,2R)-2-(ethoxycarbonyl)cyclohexan-1-aminium
Molecular FormulaC9H18NO2
InChI KeyVODUKXHGDCJEOZ-HTQZYQBOSA-O
SMILESCCOC(=O)[C@@H]1CCCC[C@H]1[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration Argentometric>=97.5 %
Appearance (Form)Granular powder
Melting point152°C to 156°C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only