cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%
cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%
cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%
Thermo Scientific Chemicals

cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%

CAS: 24947-68-0 | C7H16NO | 130.21 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
產品號碼 265880010
價格 (TWD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
規格
Assay Percent Range98.5% min. (Argentometry)
CAS24947-68-0
Chemical Name or Materialcis-2-Aminomethyl-1-cyclohexanol hydrochloride
ColorWhite
Formula Weight165.66
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only