cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%
cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%
cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%
Thermo Scientific Chemicals

cis-2-Aminomethyl-1-cyclohexanol hydrochloride, 99%

CAS: 24947-68-0 | C7H16NO | 130.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 265882500
Price (USD)
-
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS24947-68-0
IUPAC Name[(1S,2S)-2-hydroxycyclohexyl]methanaminium
Molecular FormulaC7H16NO
InChI KeyJGKFBZBVCAWDFD-BQBZGAKWSA-O
SMILES[NH3+]C[C@@H]1CCCC[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Appearance (Color)White
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only