Product Image
Thermo Scientific™
Nalgene™ Polypropylene, End Caps, mini
For Use With: Nalgene Sanitary Carboys
Have Questions?
카탈로그 번호수량
2665-0075Each
카탈로그 번호 2665-0075
제품 가격(KRW)
-
수량:
Each
Securely close off 3/4 in. or 3 in. sanitary ports on carboys with Thermo Scientific™ Nalgene™ Polypropylene End Caps. These are reliable, repeatable seals when used in combination with a silicone gasket and true union clamp. A groove on underside of end cap accepts standard sanitary gaskets for sealing the systems.
  • Seals sanitary ports on Nalgene Sanitary Carboys (No. 2630 and 2640)
  • USP Class VI
  • Autoclavable
사양
마개 크기19 mm
설명0.75 in. Mini (19mm)
직경(미터법) 내부 튜브16 mm
용도(장비)Nalgene Sanitary Carboys
수량Each
물질Polypropylene
제품 유형Cap
Unit SizeEach