Product Image
Thermo Scientific™
Nalgene™ Polypropylene, End Caps, tri
For Use With: Nalgene Sanitary Carboys
Have Questions?
카탈로그 번호수량
2665-0300Each
카탈로그 번호 2665-0300
제품 가격(KRW)
-
수량:
Each
Securely close off 3/4 in. or 3 in. sanitary ports on carboys with Thermo Scientific™ Nalgene™ Polypropylene End Caps. These are reliable, repeatable seals when used in combination with a silicone gasket and true union clamp. A groove on underside of end cap accepts standard sanitary gaskets for sealing the systems.
  • Seals sanitary ports on Nalgene Sanitary Carboys (No. 2630 and 2640)
  • USP Class VI
  • Autoclavable
사양
마개 크기76 mm
설명3 in. Tri (76mm)
용도(장비)Nalgene Sanitary Carboys
수량Each
물질Polypropylene
제품 유형Cap
Unit SizeEach