Pierce™ Sulfo-NHS-Acetate
Thermo Scientific™

Pierce™ Sulfo-NHS-Acetate

Thermo Scientific Pierce Sulfo-NHS-Acetate is a small compound used for blocking primary amines (e.g., lysine side chains). This compound forms자세히 알아보기
Have Questions?
카탈로그 번호수량
26777100 mg
카탈로그 번호 26777
제품 가격(KRW)
268,000
온라인 행사
Ends: 30-Sep-2026
315,000
할인액 47,000 (15%)
Each
수량:
100 mg
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
268,000
온라인 행사
Ends: 30-Sep-2026
315,000
할인액 47,000 (15%)
Each
Thermo Scientific Pierce Sulfo-NHS-Acetate is a small compound used for blocking primary amines (e.g., lysine side chains). This compound forms stable, covalent amide bonds with amine groups to prevent specific interactions or conjugations.

Sulfo-NHS-Acetate is typically used to prevent polymerization when performing protein crosslinking reactions and when conjugating peptides to carrier proteins for immunogen production. Blocking amines on the peptide allows directed conjugation of carboxylic acids on the peptide to primary amines on the protein using EDC.

Features of Sulfo-NHS-Acetate:

• Irreversibly block primary amines
• Perform reactions in standard non-amine-containing buffers at pH 7 or greater
• Prevent polymerization when performing protein crosslinking reactions
For Research Use Only. Not for use in diagnostic procedures.
사양
형태Powder
라벨 유형Blocking Groups
라벨 또는 염료Unreactive Group
제품라인Pierce
수량100 mg
반응성 부분Active Ester, Succinimidyl Ester, NHS Ester
용해도DMSO, Water, DMF
화학물질 반응성Amine
제품 유형Sulfo-NHS-Acetate
스페이서Short (<10 Å)
Unit SizeEach
구성 및 보관
Upon receipt store desiccated at -20° C. Product is shipped at ambient temperature.

자주 묻는 질문(FAQ)

Can you provide the shelf-life for Sulfo-NHS-Acetate?

Sulfo-NHS-Acetate is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.