Chlortetracycline hydrochloride, can be used as secondary standard
Chlortetracycline hydrochloride, can be used as secondary standard
Chlortetracycline hydrochloride, can be used as secondary standard
Thermo Scientific Chemicals

Chlortetracycline hydrochloride, can be used as secondary standard

Chlortetracycline hydrochloride, CAS # 64-72-2, is a broad-spectrum antibiotic of the tetracycline family
製品番号(カタログ番号)数量パッケージ
2682400252.5 gGlass Bottle
268245000500 mgGlass Bottle
製品番号(カタログ番号) 268240025
価格(JPY)
-
数量:
2.5 g
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS64-72-2
IUPAC Namehydrogen (2Z,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-7-chloro-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Molecular FormulaC22H24Cl2N2O8
InChI KeyRRXOSDMYCQSQHB-MRYPCDLHSA-N
SMILES[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC(Cl)=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)C(=C(N)O)C1=O
さらに表示
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Purity>=80 % (as is, calculated)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Chlortetracycline hydrochloride is a broad-spectrum antibiotic of the tetracycline family
• Receives its name from a 7-chloro substitution on one of the rings of its parent structure, tetracycline
• A bacteriostatic that inhibits the 30S ribosomal subunit and prevents peptide formation
• Can work as an antiprotozoal and antifungal
Applications
• Chlortetracycline is suitable for use in biochemistry, microbiology, in vivo research, and cell culture
• Intended for research use only
RUO – Research Use Only
Literature references
Chen, C.; Ray, P.; Knowlton, K.F.; Pruden, A.; Xia, K. Effect of composting and soil type on dissipation of veterinary antibiotics in land-applied manures. Chemosphere. 2018, 196:270-279.
Weinroth, M.D.; Scott, H.M.; Norby, B.; Loneragan, G.H.; Noyes, N.R.; Rovira, P.; Doster, E.; Yang, X.; Woerner, D.R.; Morley, P.S.; Belk, K.E. Effects of Ceftiofur and Chlortetracycline on the Resistomes of Feedlot Cattle. Appl Environ Microbiol. 2018, 84(13), e00610-18.