Chlortetracycline hydrochloride, can be used as secondary standard
Chlortetracycline hydrochloride, can be used as secondary standard
Chlortetracycline hydrochloride, can be used as secondary standard
Thermo Scientific Chemicals

Chlortetracycline hydrochloride, can be used as secondary standard

Chlortetracycline hydrochloride, CAS # 64-72-2, is a broad-spectrum antibiotic of the tetracycline family
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityPackaging
268245000500 mgGlass Bottle
2682400252.5 gGlass Bottle
Catalog number 268245000
Price (EUR)
70,90
Each
Quantity:
500 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
70,90
Each
Chemical Identifiers
CAS64-72-2
IUPAC Namehydrogen (2Z,4S,4aS,5aS,6S,12aS)-2-[amino(hydroxy)methylidene]-7-chloro-4-(dimethylamino)-6,10,11,12a-tetrahydroxy-6-methyl-1,2,3,4,4a,5,5a,6,12,12a-decahydrotetracene-1,3,12-trione chloride
Molecular FormulaC22H24Cl2N2O8
InChI KeyRRXOSDMYCQSQHB-MRYPCDLHSA-N
SMILES[H+].[Cl-].CN(C)[C@H]1[C@@H]2C[C@H]3C(=C(O)C4=C(O)C=CC(Cl)=C4[C@@]3(C)O)C(=O)[C@]2(O)C(=O)C(=C(N)O)C1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Purity>=80 % (as is, calculated)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General description
• Chlortetracycline hydrochloride is a broad-spectrum antibiotic of the tetracycline family
• Receives its name from a 7-chloro substitution on one of the rings of its parent structure, tetracycline
• A bacteriostatic that inhibits the 30S ribosomal subunit and prevents peptide formation
• Can work as an antiprotozoal and antifungal
Applications
• Chlortetracycline is suitable for use in biochemistry, microbiology, in vivo research, and cell culture
• Intended for research use only
RUO – Research Use Only
Literature references
Chen, C.; Ray, P.; Knowlton, K.F.; Pruden, A.; Xia, K. Effect of composting and soil type on dissipation of veterinary antibiotics in land-applied manures. Chemosphere. 2018, 196:270-279.
Weinroth, M.D.; Scott, H.M.; Norby, B.; Loneragan, G.H.; Noyes, N.R.; Rovira, P.; Doster, E.; Yang, X.; Woerner, D.R.; Morley, P.S.; Belk, K.E. Effects of Ceftiofur and Chlortetracycline on the Resistomes of Feedlot Cattle. Appl Environ Microbiol. 2018, 84(13), e00610-18.