Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph.
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph.
Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph.
Thermo Scientific Chemicals

Acetonitrile, 99.9%, for HPLC, far UV, conform Eur.Ph.

Green features
CAS: 75-05-8 | C2H3N | 41.053 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 268260010
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
156.65
Online Exclusive
174.00
Save 17.35 (10%)
Each
Chemical Identifiers
CAS75-05-8
IUPAC Nameacetonitrile
Molecular FormulaC2H3N
InChI KeyWEVYAHXRMPXWCK-UHFFFAOYSA-N
SMILESCC#N
View more
SpecificationsSpecification SheetSpecification Sheet
UVat 200 nm A: =<0.1
UVat 210 nm A: =<0.035
UVat 230 nm A: =<0.01
UVat 250 nm A: =<0.005
Color scale=<10 APHA
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
RUO – Research Use Only