Hexanes, for HPLC, contains ca. 95% n-Hexane
Hexanes, for HPLC, contains ca. 95% n-Hexane
Hexanes, for HPLC, contains ca. 95% n-Hexane
Hexanes, for HPLC, contains ca. 95% n-Hexane
Thermo Scientific Chemicals

Hexanes, for HPLC, contains ca. 95% n-Hexane

CAS: 92112-69-1 | C6H14
Have Questions?
Catalog NumberQuantityPackaging
2683600252.5 LGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 268360025
Price (USD)
-
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS92112-69-1
IUPAC Namehexane
Molecular FormulaC6H14
InChI KeyVLKZOEOYAKHREP-UHFFFAOYSA-N
SMILESCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Acidity=<0.00015 meq/g
UVat 200 nm A: =<0.70
UVat 220 nm A: =<0.07
UV1 cm cell vs HPLC-grade water
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For HPLC
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only