1-Pentene, 97%
1-Pentene, 97%
1-Pentene, 97%
Thermo Scientific Chemicals

1-Pentene, 97%

CAS: 109-67-1 | C5H10 | 70.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number 268660025
Price (USD)
623.65
Special offer
Online exclusive
Ends: 28-Dec-2026
733.00
Save 109.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
623.65
Special offer
Online exclusive
Ends: 28-Dec-2026
733.00
Save 109.35
Each
Chemical Identifiers
CAS109-67-1
IUPAC Namepent-1-ene
Molecular FormulaC5H10
InChI KeyYWAKXRMUMFPDSH-UHFFFAOYSA-N
SMILESCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
Refractive index1.3704 to 1.3724 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only