2,3-Lutidine, 98+%
2,3-Lutidine, 98+%
2,3-Lutidine, 98+%
Thermo Scientific Chemicals

2,3-Lutidine, 98+%

CAS: 583-61-9 | C7H9N | 107.16 g/mol
Have Questions?
Catalog NumberQuantity
26876025025 mL
Catalog number 268760250
Price (USD)
-
Quantity:
25 mL
Request bulk or custom format
Chemical Identifiers
CAS583-61-9
IUPAC Name2,3-dimethylpyridine
Molecular FormulaC7H9N
InChI KeyHPYNZHMRTTWQTB-UHFFFAOYSA-N
SMILESCC1=CC=CN=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Water=<0.5 %
GC=<0.7 % alpha-Picoline
GC>=98.0 %
GCImpurities profile
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only