Tetrabutylammonium dihydrogentrifluoride, 50-55 wt.% solution in 1,2-dichloroethane
Tetrabutylammonium dihydrogentrifluoride, 50-55 wt.% solution in 1,2-dichloroethane
Tetrabutylammonium dihydrogentrifluoride, 50-55 wt.% solution in 1,2-dichloroethane
Thermo Scientific Chemicals

Tetrabutylammonium dihydrogentrifluoride, 50-55 wt.% solution in 1,2-dichloroethane

CAS: 99337-56-1 | C16H38F3N | 301.48 g/mol
Have Questions?
Catalog NumberQuantity
2687800252.5 g
Catalog number 268780025
Price (USD)
-
Quantity:
2.5 g
Request bulk or custom format
Specifications
CAS99337-56-1
CAS107-06-2
Chemical Name or MaterialTetrabutylammonium dihydrogentrifluoride
ColorYellow to Brown
Concentration50 to 55% (NMR)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Phase transfer catalyst
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only