Lithium diisopropylamide, 2M sol. in THF/n-heptane/ethylbenzene, AcroSeal™
Lithium diisopropylamide, 2M sol. in THF/n-heptane/ethylbenzene, AcroSeal™
Lithium diisopropylamide, 2M sol. in THF/n-heptane/ethylbenzene, AcroSeal™
Acros Organics

Lithium diisopropylamide, 2M sol. in THF/n-heptane/ethylbenzene, AcroSeal™

CAS: 4111-54-0 | C6H14LiN | 107.125 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 268831000
HSN 29241900
Price (INR)
14,470.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4111-54-0
IUPAC Namelithium(1+) bis(propan-2-yl)azanide
Molecular FormulaC6H14LiN
InChI KeyZCSHNCUQKCANBX-UHFFFAOYSA-N
SMILES[Li+].CC(C)[N-]C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark yellow to orange to dark red-brown
AppearanceIf precipitation is present, gently mix
Titration with HClO424.6 to 30.0 %
Appearance (Form)Clear to slightly hazy solution
Appearanceatmosphere
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only