Nalgene™ Polypropylene, End Cap
Product Image
Thermo Scientific™
Nalgene™ Polypropylene, End Cap
For Use With: Nalgene Carboy 2630 and 2261
Have Questions?
Catalog NumberQuantity
2689-2075
Catalog number 2689-2075
Price (USD)
96.10
Case of 4
Add to cart
Price (USD)
96.10
Case of 4
Add to cart
Sanitary ports for easy fill/ing and dispensing. Thermo Scientific™ Nalgene™ End Caps in polycarbonate or polypropylene to provide operations. Caps also can be securely mounted to 3 in. sanitary flange for applications used with Nalgene™ Sanitary Carboys.
  • Autoclavable
  • Compatible with Nalgene Sanitary Carboys (Cat. Nos. 2630 and 2261)
Specifications
DescriptionTwo 0.75 in. (19mm) sanitary ports, polypropylene
Diameter (English) Inner Tubing0.75 in.
Diameter (Metric) Inner Tubing19 mm
For Use With (Equipment)Nalgene Sanitary Carboys
No. per Case4/Cs.
No. per Pack1 Ea.
MaterialPolypropylene
Product TypeEnd Cap
Unit SizeCase of 4