SPECTRONIC™ 200 Spectrophotometer Accessories
SPECTRONIC™ 200 Spectrophotometer Accessories
Imagem ilustrativa. O produto pode variar
SPECTRONIC™ 200 Spectrophotometer Accessories
SPECTRONIC™ 200 Spectrophotometer Accessories
SPECTRONIC™ 200 Spectrophotometer Accessories
Thermo Scientific™
SPECTRONIC™ 200 Spectrophotometer Accessories
Accessories for Thermo Scientific™ SPECTRONIC 200 Spectrophotometer
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoType
269-268100UV-Vis Spectrum Poster
222-265600Cuvette Rack
840-250500Cell Holder
840-250600Cylindrical Cell Holder
840-250700Replacement Sample Compartment
840-253100SPECTRONIC Standards 2 Kit
840-234000Thermal Printer
Número do catálogo 269-268100
Preço (BRL)
-
Type:
UV-Vis Spectrum Poster
Accessories for Thermo Scientific™ SPECTRONIC 200 Spectrophotometer
The SPECTRONIC 200 is a rugged, simple-to-use spectrophotometer ideal for routine applications. On-board software, controlled from an intuitive navigation pad using a full color screen, delivers capabilities beyond expectations.
Rugged, Practical Design
  • With no internal parts, there's nothing to wear out
  • Sample compartment lifts out for easy cleaning
  • Polymer construction resists corrosion from acidic or oxidizing solutions
  • Navigation pad and on-screen menus are second nature to any modern user
  • Compact footprint saves space in your lab
  • Cuvette racks on both sides of the sample compartment help you keep track of samples
  • No computer or serial cable required; Just plug it in and you're ready to measure
The Perfect Tool for Routine Spectroscopic Measurements
  • Large, bright, color LCD adjusts to the perfect angle for any technician to view
  • Standard sample compartment accommodates both 10mm square cuvettes and test tubes up to 13mm (0.5 inch) diameter
  • Room light immune optical system lets you measure tall test tubes with the lid open
  • Analyzer mode allows you to program standard methods
  • Screen-print feature gives a permanent record of your data
  • Quantitative analysis with either a factor or up to four standards offers confidence
  • Available Thermo Scientific VISIONlite5 software, for your computer, controls the instrument and provides data manipulation, storage and report printing options
Not ready to part with your old Thermo Scientific Spec 20 or GENESYS 20
  • The SPECTRONIC 200 will emulate the interface and controls on your old instrument to integrate seamlessly into your laboratory
Check the performance and accuracy of your spectrophotometer right in your own laboratory using the Thermo Scientific™ SPECTRONIC™ Spectrophotometer Standards. This kit is as easy to use as a standard cuvette and contains:
  • Calibrated neutral density glass standards for checking photometric accuracy at nominal values of 0.3A, 0.5A, 1A and 1.5A
  • Calibrated standard for checking wavelength accuracy at nominal wavelengths on 400nm, 522nm and 777nm
  • Filter for checking stray light in UV-Vis instruments at 220nm
  • Filter for checking stray light in Vis instruments at 400nm
  • Set of 7 glass pieces mounted in metal frames sized to fit standard square cuvette holders
Four calibrated neutral density glass standards for checking photometric accuracy at nominal values of 0.3A, 0.5A, 1A and 1.5A Standards are calibrated at 440.0n, 465.0nm, 546.1nm, 590.0nm and 635.0nm
One calibrated standard for checking wavelength accuracy at nominal wavelengths on 400nm, 522nm and 777nm
The symmetry of the peaks means that tests of wavelength accuracy are independent of the spectral bandwidth (between 1nm and 10nm) of the instrument being tested
1 filter for checking stray light in UV-Vis instruments at 220nm
1 filter for checking stray light in Vis instruments at 400nm
Each set of SPECTRONIC Standards is individually tested and certified to be accurate, with values traceable to the National Institute of Standards and Technology (NIST) or other Primary Standards
Filters are supplied in a tough plastic storage container
Recalibration service is available from Thermo Fisher Scientific. Recommended recalibration interval is two years
Note: Stray light filters should block light completely at the measurement wavelength and are therefore not something that can be calibrated.
Especificações
For Use With (Equipment)SPECTRONIC 200/200E Spectrophotometers
TypeUV-Vis Spectrum Poster
Unit SizeEach