Acetylsalicylsalicylic acid, 97%
Acetylsalicylsalicylic acid, 97%
Acetylsalicylsalicylic acid, 97%
Thermo Scientific Chemicals

Acetylsalicylsalicylic acid, 97%

CAS: 530-75-6 | C16H12O6 | 300.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Catalog number 269352500
Price (USD)
222.65
Online exclusive
247.00
Save 24.35
Each
Quantity:
250 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
222.65
Online exclusive
247.00
Save 24.35
Each
Chemical Identifiers
CAS530-75-6
IUPAC Name2-[2-(acetyloxy)benzoyloxy]benzoic acid
Molecular FormulaC16H12O6
InChI KeyDDSFKIFGAPZBSR-UHFFFAOYSA-N
SMILESCC(=O)OC1=CC=CC=C1C(=O)OC1=CC=CC=C1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Water=<0.5 %
Melting point145°C to 160°C
HPLC>=96.0 % (As is) Calculated
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only