Ethyl 3-ethoxypropionate, 99+%, pure, stabilized
Ethyl 3-ethoxypropionate, 99+%, pure, stabilized
Ethyl 3-ethoxypropionate, 99+%, pure, stabilized
Thermo Scientific Chemicals

Ethyl 3-ethoxypropionate, 99+%, pure, stabilized

CAS: 763-69-9 | C7H14O3 | 146.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
10 L
Packaging:
View all
Glass bottle
Catalog number 269490025
Price (USD)
117.65
Online exclusive
130.00
Save 12.35
Each
Quantity:
2.5 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
117.65
Online exclusive
130.00
Save 12.35
Each
Chemical Identifiers
CAS763-69-9
IUPAC Nameethyl 3-ethoxypropanoate
Molecular FormulaC7H14O3
InChI KeyBHXIWUJLHYHGSJ-UHFFFAOYSA-N
SMILESCCOCCC(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Infrared spectrumConforms
Color scale=<15 APHA
Refractive index1.4060 to 1.4080 (20°C, 589 nm)
Stabilizer50 to 100 ppm BHT
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only