1-Bromo-3,4-dichlorobenzene, 98+%
1-Bromo-3,4-dichlorobenzene, 98+%
1-Bromo-3,4-dichlorobenzene, 98+%
Thermo Scientific Chemicals

1-Bromo-3,4-dichlorobenzene, 98+%

CAS: 18282-59-2 | C6H3BrCl2 | 225.89 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 269811000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS18282-59-2
IUPAC Name4-bromo-1,2-dichlorobenzene
Molecular FormulaC6H3BrCl2
InChI KeyCFPZDVAZISWERM-UHFFFAOYSA-N
SMILESClC1=CC=C(Br)C=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid after melting
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.5986 to 1.6006 (20°C, 589 nm)
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only