TRACE™ GC Columns for Drugs of Abuse
Thermo Scientific™
TRACE™ GC Columns for Drugs of Abuse
Analyze common drugs of abuse with these GC columns and achieve high performance, sensitivity and reproducibility.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (Metric)
26AF130P0.25 mm
26AC497P0.2 mm
Catalog number 26AF130P
Price (TWD)
-
Diameter (Metric):
0.25 mm
Analyze amphetamines, codeine, morphine, and THC with high performance and reproducibility using Thermo Scientific™ TRACE™ GC Columns for Drugs of Abuse.
Tailored Specifically for Drugs of Abuse:
  • TRACE TR-DoA 5MS: Widely used for the analysis and determination of a range of toxicological target compounds including amphetamines, codeine, and morphine
  • TRACE TR-DoA 35MS: Ideal for the determination of carboxy-THC because of its higher polarity, making it the recommended column for use in drug testing labs for the confirmation of THC
Excellent Quality and Reproducibility:
  • Low bleed even at elevated temperatures
  • Outstanding robustness for difficult separations
  • Reliable and reproducible performance
Specifications
Diameter (Metric)0.25 mm
For Use With (Application)GC/MS for Amphetamines, Codeine, and Morphine
Max. Temperature (Metric)360/370°C
Diameter (Metric) Inner0.25 mm
Film Thickness0.25 μm
Length (Metric)15 m
Product LineTRACE
Stationary PhaseTrace™ TR-DoA 35 MS
USP TypeG42
Unit SizePack of 1