Thermo Scientific™
TraceGOLD™ TG-WaxMT Metal GC Columns
Catalog number: 26M882860
Thermo Scientific™
TraceGOLD™ TG-WaxMT Metal GC Columns
Catalog number: 26M882860
Ensure excellent column-to-column reproducibility using the Thermo Scientific™ TraceGOLD™ TG-WaxMT GC Columns.
Catalog Number
26M882860
Unit Size
Each
Particle Size
1 μm
Length (Metric)
15 m
Diameter (Metric)
0.53 mm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size1 μm
Temperature Range40°C°C to 260°C
Film Thickness1 μm
Length (Metric)15 m
PolarityPolar
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
50 μm
Length (Metric)
15 m
Diameter (Metric)
0.53 mm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size50 μm
Temperature Range40°C°C to 260°C
Film Thickness50 μm
Length (Metric)15 m
PolarityPolar
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
0.5 μm
Length (Metric)
30 m
Diameter (Metric)
0.25 mm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.5 μm
Temperature Range40°C°C to 260°C
Film Thickness0.5 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
0.25 μm
Length (Metric)
15 m
Diameter (Metric)
0.25 mm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.25 μm
Temperature Range40°C°C to 260°C
Film Thickness0.25 μm
Length (Metric)15 m
PolarityPolar
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
0.10 μm
Length (Metric)
15 m
Diameter (Metric)
0.25 mm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.10 μm
Temperature Range40°C°C to 260°C
Film Thickness0.1 μm
Length (Metric)15 m
PolarityPolar
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
0.5 μm
Length (Metric)
30 m
Diameter (Metric)
0.53 mm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.5 μm
Temperature Range40°C°C to 260°C
Film Thickness0.5 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
0.25 μm
Length (Metric)
30 m
Diameter (Metric)
0.25 mm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.25 μm
Temperature Range40°C°C to 260°C
Film Thickness0.25 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
0.25 μm
Length (Metric)
60 m
Diameter (Metric)
0.25 mm
Price (USD)
Full specifications
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.25 μm
Temperature Range40°C°C to 260°C
Film Thickness0.25 μm
Length (Metric)60 m
PolarityPolar
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
1 μm
Length (Metric)
30 m
Diameter (Metric)
0.53 mm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size1 μm
Temperature Range40°C°C to 260°C
Film Thickness1 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
Catalog Number
Unit Size
Each
Particle Size
0.25 μm
Length (Metric)
30 m
Diameter (Metric)
0.53 mm
Price (USD)
Full specifications
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.25 μm
Temperature Range40°C°C to 260°C
Film Thickness0.25 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
Showing 10 of 11Show more
Catalog NumberSpecificationsUnit SizeParticle SizeLength (Metric)Diameter (Metric)Price (USD)
26M882860Full specifications
Each1 μm15 m0.53 mmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size1 μm
Temperature Range40°C°C to 260°C
Film Thickness1 μm
Length (Metric)15 m
PolarityPolar
Unit SizeEach
26M882130Full specifications
Each50 μm15 m0.53 mmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size50 μm
Temperature Range40°C°C to 260°C
Film Thickness50 μm
Length (Metric)15 m
PolarityPolar
Unit SizeEach
26M882230Full specifications
Each0.5 μm30 m0.25 mmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.5 μm
Temperature Range40°C°C to 260°C
Film Thickness0.5 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
26M881300Full specifications
Each0.25 μm15 m0.25 mmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.25 μm
Temperature Range40°C°C to 260°C
Film Thickness0.25 μm
Length (Metric)15 m
PolarityPolar
Unit SizeEach
26M880350Full specifications
Each0.10 μm15 m0.25 mmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.10 μm
Temperature Range40°C°C to 260°C
Film Thickness0.1 μm
Length (Metric)15 m
PolarityPolar
Unit SizeEach
26M882250Full specifications
Each0.5 μm30 m0.53 mmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.5 μm
Temperature Range40°C°C to 260°C
Film Thickness0.5 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
26M881420Full specifications
Each0.25 μm30 m0.25 mmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.25 μm
Temperature Range40°C°C to 260°C
Film Thickness0.25 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
26M881540Full specifications
Each0.25 μm60 m0.25 mmRequest A Quote
Diameter (Metric)0.25 mm
Diameter (Metric) Inner0.25 mm
Product LineTraceGOLD
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.25 μm
Temperature Range40°C°C to 260°C
Film Thickness0.25 μm
Length (Metric)60 m
PolarityPolar
Unit SizeEach
26M882880Full specifications
Each1 μm30 m0.53 mmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size1 μm
Temperature Range40°C°C to 260°C
Film Thickness1 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
26M881440Full specifications
Each0.25 μm30 m0.53 mmRequest A Quote
Diameter (Metric)0.53 mm
Diameter (Metric) Inner0.53 mm
Stationary PhaseTraceGOLD™ TG-WAXMS
USP TypeG14, G15, G16, G20, G39
Max. Temperature260°C
For Use With (Application)GC for EPA Method 603 for Acrolein/Acrylonitrile, GC for Fatty Acid Methyl Esters (FAMEs), Flavor Compounds, Essential Oils, Amines, Solvents, and Xylene Isomers
Particle Size0.25 μm
Temperature Range40°C°C to 260°C
Film Thickness0.25 μm
Length (Metric)30 m
PolarityPolar
Unit SizeEach
Showing 10 of 11Show more
Engineered for Superior Column-to-Column Reproducibility
  • Polar phase, polyethylene glycol
  • Polar-deactivated surface tightly binds polymer for excellent thermal stability
  • Resists oxidative damage, damage from strongly acidic or basic volatiles better than silicone solid phases
  • Equivalent to USP G14, G15, G20 and G39 phases
Applications:
  • Fatty acid methyl esters (FAMEs)
  • Flavor compounds
  • Essential oils
  • Amines
  • Solvents
  • Xylene isomers
  • EPA Method 603 for Acrolein/Acrylonitrile
A Leap Forward in Column Performance with Low Bleed and Superior Inertness
  • This low-bleed, stable column produces consistent results.
  • Low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Minimizing column bleed extends column lifetime, leading to higher productivity.
  • Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.
  • Superior inertness ensures excellent peak shape and sensitivity, especially for highly active or difficult-to-analyze compounds.
Recommended Use:
  • Aldehydes
  • Essential oils
  • Solvents
  • Xylenes
  • Alternative Use:
  • Acids
  • Alcohols
  • Alcohols in beverages
  • Alditol acetates (sugars)
  • Amines–Aliphatic
  • Amines–Aromatic
Similar To:
  • MXT™-Wax
  • DB-WAX
  • DB-WAXetr
  • HP-Wax
  • HP-Innowax
  • SUPELCOWAX™ 10
  • CP-Wax 52 CB