Fosforamidita rU, grado estándar, frasco vial de suero
Thermo Scientific™

Fosforamidita rU, grado estándar, frasco vial de suero

rU CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) for use in oligonucleotide synthesis. RNA phosphoramiditesMás información
Have Questions?
Número de catálogoCantidad
27-1404-072 g
Número de catálogo 27-1404-07
Precio (USD)
72,29
Each
Cantidad:
2 g
Precio (USD)
72,29
Each

rU CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups (except T and U) designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 60 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-O-TBDMSi-uridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C45H61N4O9PSi
• Base—rU
• Formula weight—861.1 g/mol
• CAS number—118362-03-1
• HPLC Purity—≥98.0%
31P NMR Purity—≥98.0%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (equipo)Sintetizadores AKTA, sintetizadores Applied Biosystems
FormularioPolvo
GradoEstándar
Tipo de ácido nucleicoARN
Tipo de nucleósidoUridina (U)
Tipo de productorU CEP
Pureza98%
Cantidad2 g
Tipo de reactivoFosforamidita
Condiciones de envíoTemperatura ambiente
Unit SizeEach
Contenido y almacenamiento
Store in freezer (at or below -20°C).