8-oxo-dG Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

8-oxo-dG Phosphoramidite, standard grade, serum vial bottle

8-oxo-dG CEP is a phosphoramidite containing an oxidized deoxyguanosine (dG), and it facilitates the investigation of the structure and activityRead more
Have Questions?
Catalog NumberQuantity
27-1424-040.25 g
Catalog number 27-1424-04
Price (TWD)
0.01
Each
Quantity:
0.25 g
Price (TWD)
0.01
Each

8-oxo-dG CEP is a phosphoramidite containing an oxidized deoxyguanosine (dG), and it facilitates the investigation of the structure and activity when incorporated into an oligonucleotide. In nature, 8-oxo-dG DNA lesions are formed with reactive oxygen species (ROS), and is removed by by the base excision repair (BER) mechanism. This modification is significant in mutagenesis, carcinogenesis, and has been proven useful as a biomarker. This product is supplied in a 5 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)‐N2-isobutyryl-8-oxo-deoxyguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C44H54N7O9P
• Base—8-oxo-dG
• Base Protection—iBu
• Formula weight—855.9 g/mol
• CAS number—143060-53-1
• HPLC Purity—≥98.0%
31P NMR Purity—≥98.0%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeDNA
Nucleoside TypeDeoxyguanosine (dG)
Product Type8-oxo-dG CEP
Purity98%
Quantity0.25 g
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).