TheraPure 2ʹ-Fluoro-Bz-A Phosphoramidite
Thermo Scientific™

TheraPure 2ʹ-Fluoro-Bz-A Phosphoramidite

2'-Fluoro-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-Fluoro protection, for use in oligonucleotide synthesis.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
27-1601-120.5 gBottle (28-400 Thread)
27-1601-021 gSerum Vial Bottle
27-1601-090.25 gAmber Bottle (20-400 Thread)
27-1601-75100 gBulk Standard
Catalog number 27-1601-12
Price (TWD)
2,197.00
Each
Quantity:
0.5 g
Format:
Bottle (28-400 Thread)
Price (TWD)
2,197.00
Each

2'-Fluoro-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-Fluoro protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, the highly electronegative fluorine (2'-Fluoro) modification adopts a more thermodynamically stable A-form helix on hybridization to its target. Showing improved affinity and some nuclease resistance (compared to 2'-OH), the 2'-F modification has been proven useful in antisense, siRNA, and aptamer applications. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in an amber bottle.

Features:

  • Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N6-benzoyl-2'-fluoroadenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
  • Molecular formula—C47H51FN7O7P
  • Base—A
  • Base Protection—Bz
  • Formula weight—875.92 g/mol
  • HPLC purity—>98%
  • 31P NMR purity—≥98%

Available with a Serum Vial, 20-400 Finish, or 28-400 Finish neck style.

  • Serum vial neck style is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.
  • 20-400 finish neck style is compatible with most Expedite synthesizers.
  • 28-400 finish neck style is compatible with most MerMade synthesizers.
For Research Use Only.
Specifications
For Use With (Equipment)MerMade Synthesizers
FormPowder
FormatBottle (28-400 Thread)
GradeStandard
Nucleic Acid TypeRNA
Nucleoside TypeAdenosine (A)
Product Type2in-Fluoro-Bz-A CEP
Purity98%
Quantity0.5 g
Reagent Features2in-Fluoro modified
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).