TheraPure™ 2'-Fluoro-Ac-C Phosphoramidite, 20-400 finish bottle
Thermo Scientific™

TheraPure™ 2'-Fluoro-Ac-C Phosphoramidite, 20-400 finish bottle

2'-Fluoro-Ac-C CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with 2'-Fluoro protection, for use in oligonucleotideRead more
Have Questions?
Catalog NumberQuantity
27-1604-090.25 g
Catalog number 27-1604-09
Price (TWD)
0.01
Each
Quantity:
0.25 g
Price (TWD)
0.01
Each

2'-Fluoro-Ac-C CEP is a phosphoramidite containing 5' DMT and the nucleoside cytidine (C) with 2'-Fluoro protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, the highly electronegative fluorine (2'-Fluoro) modification adopts a more thermodynamically stable A-form helix on hybridization to its target. Showing improved affinity and some nuclease resistance (compared to 2'-OH), the 2'-F modification has been proven useful in antisense, siRNA, and aptamer applications. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30-mL amber bottle with a 20-400 finish neck style, which is compatible with most Expedite synthesizers.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
31P NMR Purity≥98%
CAS159414-99-0
Chemical Name or Material5'-O-(4,4'-Dimethoxytrityl)-N4-acetyl-2'-fluorocytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
DescriptionBase: C
Base Protection: Ac
For Use With (Application)Oligonucleotide synthesis
For Use With (Equipment)Expedite Synthesizers
FormPowder
FormatAmber bottle (20-400 finish neck style)
GradeTheraPure™
Molecular FormulaC41H49N5FO8P
Molecular Weight (g/mol)789.8 g/mol
Nucleic Acid TypeRNA
Nucleoside TypeCytidine (C)
Product Type2in-Fluoro-Ac-C CEP
Purity>98% (HPLC)
Quantity0.25 g
Reagent Features2in-Fluoro modified, Fast deprotection
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Product LineTheraPure
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).