iPrPAC-dG Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

iPrPAC-dG Phosphoramidite, standard grade, serum vial bottle

iPrPAC-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with Isopropylphenoxyacetyl (iPrPAC) base protection, for use in oligonucleotide synthesis.
Have Questions?
Catalog NumberGrade
27-1726-04Standard
Catalog number 27-1726-04
Price (BRL)
-
Grade:
Standard

The UltraMILD monomers - phenoxyacetyl (PAC)-dA, acetyl (Ac)-dC, and iso-propyl-phenoxyacetyl (iPr-PAC)-dG - were developed to alleviate the problem of modifiers and labels not withstanding prolonged exposure to strongly alkaline deprotection conditions. Cleavage and deprotection can be achieved in 4 hours at room temperature with 0.05M potassium carbonate in methanol. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems∣AKTA Systems of oligonucleotide synthesizers.

Features:

  • Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N2-isopropylphenoxyacetyl-2'-deoxyguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
  • Molecular formula—C51H60N7O9P
  • Base—dG
  • Base Protection—iPrPAC
  • Formula weight—946.05 g/mol
  • CAS number—150065-82-0
  • HPLC Purity—≥98.0%
  • 31P NMR Purity—≥98.0%
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormatPowder
GradeStandard
Nucleic Acid TypeDNA
Nucleoside TypeDeoxyguanosine (dG)
Product TypeiPrPAC-dG CEP
Purity98%
Reagent FeaturesFast deprotection
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).