Fosforamidita Bz-dC
Thermo Scientific™

Fosforamidita Bz-dC

Bz-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Benzoyl (Bz) base protection, for useMás información
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
27-1732-055 g
27-1732-041 g
Número de catálogo 27-1732-05
Precio (CLP)
39.645
Each
Cantidad:
5 g
Precio (CLP)
39.645
Each

Bz-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 100 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N4-benzoyl-2'-deoxycytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C46H52N5O8P
• Base—dC
• Base Protection—Bz
• Formula weight—833.93 g/mol
• CAS number—102212-98-6
• HPLC Purity—≥98.0%
• 31P NMR Purity—≥98.0%

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Para utilizar con (equipo)Sintetizadores AKTA, sintetizadores Applied Biosystems
FormularioPolvo
GradoEstándar
Tipo de ácido nucleicoADN
Tipo de nucleósidoDesoxicitidina (dC)
Tipo de productoBz-dC CEP
Pureza98%
Cantidad5 g
Tipo de reactivoFosforamidita
Condiciones de envíoTemperatura ambiente
Unit SizeEach
Contenido y almacenamiento
Store in freezer (at or below -20°C).