iBu-dG Phosphoramidite, standard grade, 28-400 finish bottle
Thermo Scientific™

iBu-dG Phosphoramidite, standard grade, 28-400 finish bottle

iBu-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with isobutyryl (iBu) base protection, for use자세히 알아보기
Have Questions?
카탈로그 번호수량
27-1734-132 g
카탈로그 번호 27-1734-13
제품 가격(KRW)
1
Each
수량:
2 g
제품 가격(KRW)
1
Each

iBu-dG CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyguanosine (dG) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The 'Standard Grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 30 mL Amber bottle with a 28-400 Finish neck style, which is compatible with most MerMade Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-deoxyguanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C44H54N7O8P
• Base—dG
• Base Protection—iBu
• Formula weight—839.92 g/mol
• CAS number—93183-15-4
• HPLC Purity—≥98.0%
31P NMR Purity—≥98.0%

For Research Use Only. Not for use in diagnostic procedures.
사양
용도(장비)MerMade Synthesizers
형태Powder
등급Standard
핵산 유형DNA
뉴클레오시드 유형Deoxyguanosine (dG)
제품 유형iBu-dG CEP
순도98%
수량2 g
시약 유형Phosphoramidite
배송 조건Room Temperature
Unit SizeEach
구성 및 보관
Store in freezer (at or below -20°C).