dU Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

dU Phosphoramidite, standard grade, serum vial bottle

dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis. The incorporationRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
27-1738-03100 μM
27-1738-021 g
Catalog number 27-1738-03
Price (KRW)
1
Each
Quantity:
100 μM
Price (KRW)
1
Each

dU CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyuridine (dU) for use in oligonucleotide synthesis. The incorporation of a deoxyuridine base into a DNA sequence can be utilized to induce mutagenic effects. Enzymatically the uracil is removed, creating an abasic site at the dU position, and thus generating site-specific breaks in the oligo. This product is supplied in a 20 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyuridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C39H47N4O8P
• Base—dU
• Formula weight—730.8 g/mol
• CAS number—109389-30-2
• HPLC Purity—≥98.0%; conforms to reference
31P NMR Purity—≥98.0%; conforms to reference

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeDNA
Nucleoside TypeDeoxyuridine (dU)
Product TypedU CEP
Purity98%
Quantity100 μM
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).