dI Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

dI Phosphoramidite, standard grade, serum vial bottle

dI CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyinosine (dI) for use in oligonucleotide synthesis. The problemRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
27-1744-04250 mg
27-1744-03100 μmol
Catalog number 27-1744-04
Price (CLP)
-
Quantity:
250 mg

dI CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyinosine (dI) for use in oligonucleotide synthesis. The problem of degeneracy of the genetic code can be assuaged by utilizing universal bases, such as deoxyinosine (dI), in an oligonucleotide. Its unique structure allows for it to pair with all four bases in various ‘wobble’ structures, with an overall preferential order of base-pairing: dI-dC > dI-dA > dI-dG = dI-dT. This product is supplied in a 5 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-deoxyinosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C40H47N6O7P
• Formula weight—754.8 g/mol
• CAS number—141684-35-7
• HPLC Purity—≥98.0%; M-11 Impurity ≤0.01%
31P NMR Purity—≥98%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeDNA
Nucleoside TypeDeoxyinosine (dI)
Product TypedI Phosphoramidite
Purity98%
Quantity250 mg
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).