5me-dC Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

5me-dC Phosphoramidite, standard grade, serum vial bottle

5me-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Benzoyl (Bz) base protection, for useRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
27-1748-041 g
27-1748-055 g
Catalog number 27-1748-04
Price (KRW)
0
Each
Quantity:
1 g
Price (KRW)
0
Each

5me-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis. The 5-me-dC Phosphoramidite elicits enhanced binding and increased duplex stabilization (to dG) compared to unmethylated dC. This can be attributed to the hydrophobic nature of the methyl groups that help eliminate water molecules from the duplex. Thus, it can be utilized to improve the ability of an oligo to hybridize to its target. It has proven especially useful in oligo applications such as strong-binding PCR primers, immune stimulation (via antisense), and in methylation studies. This product is supplied in a 20 mL Amber bottle with a Serum Vial neck style, which is compatible with most ABI Systems|AKTA Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N4-Benzoyl-5-Methyl-2'-deoxycytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C47H54N5O8P
• Base—5me-dC
• Base Protection—Bz
• Formula weight—847.95 g/mol
• CAS number—105931-57-5
• HPLC Purity—>98%; M-11 Impurity ≤0.01%
31P NMR Purity—≥98%; HMT impurity (∼146ppm): ≤0.10%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeDNA
Nucleoside TypeDeoxycytidine (dC)
Product Type5me-dC CEP
Purity98%
Quantity1 g
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).