Phosphorylating Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

Phosphorylating Phosphoramidite, standard grade, serum vial bottle

Phosphorylating phosphoramidite can be utilized to achieve phosphorylation of the 5' oligo terminus (also known as Phosphate-ON or Chemical PhosphorylationRead more
Have Questions?
Catalog NumberQuantity
27-1794-040.25 g
Catalog number 27-1794-04
Price (BRL)
-
Quantity:
0.25 g

Phosphorylating phosphoramidite can be utilized to achieve phosphorylation of the 5' oligo terminus (also known as Phosphate-ON or Chemical Phosphorylation Reagent (CPR)). Chemical phosphorylation is a cost-effective substitute to enzymatic methods (via T4 kinase and ATP), allowing efficient introduction of terminal phosphate groups. Oligos with a 5'-phosphate are conducive to a number of various applications utilizing the ligation of oligos to each other, such as NGS adapters, cloning, and gene construction. This product is supplied in a 5 mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers of oligonucleotide synthesizers.

Features:
• Full chemical name—2-[2-(4,4-Dimethoxytrityloxy)ethylsulfonyl]ethyl-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C34H45F3N2O7PS
• Formula weight—656.66 g/mol
• HPLC Purity—≥95.0%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeNone (Modifier)
Nucleoside TypeNone (Modifier)
Product TypePhosphorylating CEP
Purity95%
Quantity0.25 g
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).