Phosphorylating Phosphoramidite, standard grade, 20-400 finish bottle
Thermo Scientific™

Phosphorylating Phosphoramidite, standard grade, 20-400 finish bottle

Phosphorylating phosphoramidite can be utilized to achieve phosphorylation of the 5' oligo terminus (also known as Phosphate-ON or Chemical PhosphorylationRead more
Have Questions?
Catalog NumberQuantity
27-1794-09100 μmol
Catalog number 27-1794-09
Price (CLP)
-
Quantity:
100 μmol

Phosphorylating phosphoramidite can be utilized to achieve phosphorylation of the 5' oligo terminus (also known as Phosphate-ON or Chemical Phosphorylation Reagent (CPR)). Chemical phosphorylation is a cost-effective substitute to enzymatic methods (via T4 kinase and ATP), allowing efficient introduction of terminal phosphate groups. Oligos with a 5'-phosphate are conducive to a number of various applications utilizing the ligation of oligos to each other, such as NGS adapters, cloning, and gene construction. This product is supplied in a 30 mL amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—2-[2-(4,4-Dimethoxytrityloxy)ethylsulfonyl]ethyl-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C34H45F3N2O7PS
• Formula weight—656.66 g/mol
• HPLC Purity—≥95.0%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Expedite Synthesizers
FormPowder
GradeStandard
Nucleic Acid TypeNone (Modifier)
Nucleoside TypeNone (Modifier)
Product TypePhosphorylating CEP
Purity95%
Quantity100 μmol
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).