2'-OMe-Bz-A Phosphoramidite, standard grade, serum vial bottle
Thermo Scientific™

2'-OMe-Bz-A Phosphoramidite, standard grade, serum vial bottle

2'-OMe-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-O-methyl protection, for use in oligonucleotideRead more
Have Questions?
Catalog NumberQuantity
27-1842-021 g
Catalog number 27-1842-02
Price (EUR)
21,02
Each
Quantity:
1 g
Price (EUR)
21,02
Each
2'-OMe-Bz-A CEP is a phosphoramidite containing 5' DMT and the nucleoside adenosine (A) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The 'standard grade' marking denotes a baseline level of purity verified with analytical methods, and proven to be suitable for most oligo synthesis requirements. This product is supplied in a 20-mL amber bottle with a serum vial neck style, which is compatible with most Applied Biosystems synthesizers and AKTA synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N6-benzoyl-2'-O-methyl-adenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C48H54N7O8P
• Base—A
• Base Protection—Bz
• Formula weight—887.97 g/mol
• CAS number—110782-31-5
• HPLC purity—≥98.0%; Sum of non-primary peaks 140-152ppm: ≤0.5%
31P NMR purity—≥98.0%

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)AKTA Synthesizers, Applied Biosystems Synthesizers
FormatPowder
GradeStandard
Nucleic Acid TypeRNA
Nucleoside TypeAdenosine (A)
Product Type2'-OMe-Bz-A CEP
Purity98%
Quantity1 g
Reagent Features2'-OMe modified
Reagent TypePhosphoramidite
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store in freezer (at or below -20°C).