Fosforamidita iBu-rG TheraPure™, frasco de terminado 28-400
Thermo Scientific™

Fosforamidita iBu-rG TheraPure™, frasco de terminado 28-400

iBu-rG CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with isobutyryl (iBu) base protection, for useMás información
Have Questions?
Número de catálogoCantidad
27-1906-132 g
Número de catálogo 27-1906-13
Precio (CLP)
51.677
Each
Cantidad:
2 g
Precio (CLP)
51.677
Each

iBu-rG CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30 mL Amber bottle with a 28-400 Finish neck style, which is compatible with most MerMade Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-[(tert-butyl)dimethylsilyl]-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C50H68N7O9PSi
• Base Protection—iBu
• Formula weight—970.1 g/mol
• CAS number—147201-04-5
• HPLC Purity—≥99%; M-11 Impurity: ≤0.01%
31P NMR Purity—≥99%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%; No single PIII impurity >0.3%

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (equipo)Sintetizadores MerMade
FormularioPolvo
GradoTheraPure™
Tipo de ácido nucleicoARN
Tipo de nucleósidoGuanosina (G)
Tipo de productoCEP iBu-rG
Pureza99%
Cantidad2 g
Tipo de reactivoFosforamidita
Condiciones de envíoTemperatura ambiente
Línea de productosTheraPure
Unit SizeEach
Contenido y almacenamiento
Store in freezer (at or below -20°C).