TheraPure™ iBu-rG Phosphoramidite, wide mouth bottle
Thermo Scientific™

TheraPure™ iBu-rG Phosphoramidite, wide mouth bottle

iBu-rG CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis.
Have Questions?
카탈로그 번호수량
27-1906-5050 g
카탈로그 번호 27-1906-50
제품 가격(KRW)
1,441,000
Each
수량:
50 g
제품 가격(KRW)
1,441,000
Each

iBu-rG CEP is a phosphoramidite containing 5' DMT and the nucleoside guanosine (G) with isobutyryl (iBu) base protection, for use in oligonucleotide synthesis. RNA phosphoramidites contain a 2'-OH protecting group, tBDMS (tert-butyl dimethylsilyl). Each base has unique protecting groups designed specifically to protect the RNA during the chemical synthesis process, and produce the highest amount of full length product. Alternate base protection options are also available to allow for faster deprotection and longer oligonucleotide synthesis. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 250 mL Opaque bottle with a Wide Mouth neck style, which is compatible with most None (Bulk) of oligonucleotide synthesizers.

Features:
• HPLC Purity—≥99%; M-11 Impurity: ≤0.01%
• 31P NMR Purity—≥99%; Sum of non-primary peaks in 140-152 ppm range: ≤0.5%; No single PIII impurity >0.3%

For Research Use Only. Not for use in diagnostic procedures.
사양
31P NMR 순도≥99%
CAS147201-04-5
화학물질 이름 또는 재질5'-O-(4,4'-Dimethoxytrityl)-N2-isobutyryl-2'-O-[(tert-butyl)dimethylsilyl]-guanosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
설명Base: rG
Base Protection: iBu
용도(애플리케이션)Oligonucleotide synthesis
형태Powder
형식Opaque bottle, wide-mouth
등급TheraPure™
분자식C50H68N7O9Psi
분자량970.1 g/mol
핵산 유형RNA
뉴클레오시드 유형Guanosine (G)
제품 유형iBu-rG CEP
순도≥99%
수량50 g
시약 유형Phosphoramidite
배송 조건Room Temperature
제품라인TheraPure
Unit SizeEach
구성 및 보관
Store in freezer (at or below -20°C).