Product Image
Thermo Scientific™

TheraPure™ Bz-dA Phosphoramidite, 28-400 finish bottle

Bz-dA CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyadenosine (dA) with Benzoyl (Bz) base protection, for use자세히 알아보기
Have Questions?
카탈로그 번호수량
27-2030-1410 g
카탈로그 번호 27-2030-14
제품 가격(KRW)
115,000
Each
수량:
10 g
제품 가격(KRW)
115,000
Each

Bz-dA CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxyadenosine (dA) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis. DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The TheraPure trademark denotes a highest level of phosphoramidite purity, developed with the highest stringency of of high definiteion analytics and tightly controlled impurity profiles. Ideal for any therapeutic application. This product is supplied in a 250 mL Amber bottle with a 28-400 Finish neck style, which is compatible with most MerMade Systems of oligonucleotide synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N6-benzoyl-2'-deoxyadenosine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C47H52N7O7P
• Base—dA
• Base Protection—Bz
• Formula weight—857.95 g/mol
• CAS number—98796-53-3
• HPLC Purity—≥99%; M-11 Impurity ≤0.01%; Single critical impurity ≤0.3%; Presence of dC, dG, or T impurity ≤0.03%
31P NMR Purity—≥99%; Sum of non-primary peaks at 140-152 ppm: ≤0.3%; No peak(s) above 170 ppm; HMT impurity (∼146 ppm): ≤0.10%

For Research Use Only. Not for use in diagnostic procedures.
사양
용도(장비)MerMade Synthesizers
형태Powder
등급TheraPure™
핵산 유형DNA
뉴클레오시드 유형Deoxyadenosine (dA)
제품 유형Bz-dA CEP
순도99%
수량10 g
시약 유형Phosphoramidite
배송 조건Room Temperature
제품라인TheraPure
Unit SizeEach
구성 및 보관
Store in freezer (at or below -20°C).