Product Image
Thermo Scientific™

TheraPure™ Bz-dC Phosphoramidite, 20-400 finish bottle

Bz-dC CEP is a phosphoramidite containing 5' DMT and the nucleoside deoxycytidine (dC) with Benzoyl (Bz) base protection, for use in oligonucleotide synthesis.
Have Questions?
Número de catálogoCantidad
27-2032-841 frasco
Número de catálogo 27-2032-84
Precio (USD)
33,88
Each
Cantidad:
1 frasco
Precio (USD)
33,88
Each

DNA phosphoramidites are the building blocks for synthesis of DNA or DNA containing oligonucleotides. Each base has unique protecting groups designed specifically to protect the DNA during the chemical synthesis process, and produce the highest amount of full length product. Alternative protecting groups on dC (acetyl) and dG (DMF) allow for a faster deprotection of the oligonucleotide. The TheraPure trademark denotes a highest level of phosphoramidite purity, developed with the highest stringency of of high definition analytics and tightly controlled impurity profiles. Ideal for any therapeutic application. This product is supplied in a 60 mL Amber bottle with a 20-400 Finish neck style, which is compatible with most Expedite Systems of oligonucleotide synthesizers.

Features:

  • Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-N4-benzoyl-2'-deoxycytidine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
  • Molecular formula—C46H52N5O8P
  • Base—dC
  • Base Protection—Bz
  • Formula weight—833.93 g/mol
  • CAS number—102212-98-6
  • HPLC Purity—≥99%; M-11 Impurity ≤0.01%; Single critical impurity ≤0.3%; Presence of dA, dG, or T impurity ≤0.03%
  • 31P NMR Purity—≥99%; Sum of non-primary peaks at 140-152 ppm: ≤0.3%; No peak(s) above 170 ppm; HMT impurity (∼146ppm): ≤0.10%

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.

Especificaciones
Para utilizar con (equipo)Sintetizadores Expedite
FormatoPolvo
GradoTheraPure™
Tipo de ácido nucleicoADN
Tipo de nucleósidoDesoxicitidina (dC)
Tipo de productoBz-dC CEP
Pureza99%
Cantidad1 frasco
Tipo de reactivoFosforamidita
Condiciones de envíoTemperatura ambiente
Línea de productosTheraPure
Unit SizeEach
Contenido y almacenamiento
Store in freezer (at or below -20°C).