Fosforamidita 2'-OMe-U TheraPure™, frasco de acabado 20-400
Thermo Scientific™

Fosforamidita 2'-OMe-U TheraPure™, frasco de acabado 20-400

2'-OMe-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-O-methyl protection, for use in oligonucleotideMás información
Have Questions?
Número de catálogoCantidad
27-2044-101 g
Número de catálogo 27-2044-10
Precio (CLP)
26.434
Each
Cantidad:
1 g
Precio (CLP)
26.434
Each

2'-OMe-U CEP is a phosphoramidite containing 5' DMT and the nucleoside uridine (U) with 2'-O-methyl protection, for use in oligonucleotide synthesis. Located in the ideal oligo position, 2'-O-methyl (OMe) oligo modifications display nuclease resistance, fewer off-target effects, and slightly increased hybridization affinities. These phosphoramidites are commonly used for oligos designed for antisense, siRNA, aptamer, or gRNA-based research. The TheraPure trademark denotes our highest level of phosphoramidite purity, developed with the greatest stringency of high definition analytics and tightly controlled impurity profiles and, therefore, ideal for any therapeutic application. This product is supplied in a 30-mL amber bottle with a 20-400 finish neck style, which is compatible with most Expedite synthesizers.

Features:
• Full chemical name—5'-O-(4,4'-Dimethoxytrityl)-2'-O-methyl-uridine-3'-O-[(2-cyanoethyl)-(N,N-diisopropyl)]-phosphoramidite
• Molecular formula—C40H56N7O9P
• Base—U
• Formula weight—760.8 g/mol
• CAS number—110764-79-9
• HPLC purity—≥99%; M-11 Impurity: ≤0.01%; Single critical impurity ≤0.3%
31P NMR purity—>99%; Sum of non-primary peaks at 140-152 ppm ≤0.5%

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Para utilizar con (equipo)Sintetizadores Expedite
FormularioPolvo
GradoTheraPure™
Tipo de ácido nucleicoARN
Tipo de nucleósidoUridina (U)
Tipo de producto2in-OMe-U CEP
Pureza99%
Cantidad1 g
Características de los reactivos2in-OMe modified
Tipo de reactivoFosforamidita
Condiciones de envíoTemperatura ambiente
Línea de productosTheraPure
Unit SizeEach
Contenido y almacenamiento
Store in freezer (at or below -20°C).