3-Phenyl-2-propyn-1-ol, 98%
3-Phenyl-2-propyn-1-ol, 98%
3-Phenyl-2-propyn-1-ol, 98%
Thermo Scientific Chemicals

3-Phenyl-2-propyn-1-ol, 98%

CAS: 1504-58-1 | C9H8O | 132.16 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 270030250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS1504-58-1
IUPAC Name3-phenylprop-2-yn-1-ol
Molecular FormulaC9H8O
InChI KeyNITUNGCLDSFVDL-UHFFFAOYSA-N
SMILESOCC#CC1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
GC>=97.5 %
Appearance (Form)Liquid
Appearance (Color)Clear colorless to yellow
Refractive index1.5830 to 1.5870 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only