Thermo Scientific Chemicals

Rhodium acetate, ca. 40% Rh, brown, water soluble, Thermo Scientific Chemicals

Catalog number: 270200010
1 g, Each
Thermo Scientific Chemicals

Rhodium acetate, ca. 40% Rh, brown, water soluble, Thermo Scientific Chemicals

Catalog number: 270200010
1 g, Each
Quantity

Chemical Identifiers

CAS
42204-14-8
Appearance (Color)
Brown
Appearance (Form)
Powder
Assay
35.0 to 40.0 % (Rh)
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text