Dibenzyl azodicarboxylate, 94%
Dibenzyl azodicarboxylate, 94%
Dibenzyl azodicarboxylate, 94%
Thermo Scientific Chemicals

Dibenzyl azodicarboxylate, 94%

CAS: 2449-05-0 | C16H14N2O4 | 298.29 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 270270050
價格 (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS2449-05-0
IUPAC NameN-{[(benzyloxy)carbonyl]imino}(benzyloxy)formamide
Molecular FormulaC16H14N2O4
InChI KeyIRJKSAIGIYODAN-UHFFFAOYSA-N
SMILESO=C(OCC1=CC=CC=C1)N=NC(=O)OCC1=CC=CC=C1
檢視更多
規格Specification Sheet規格表
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point44°C to 52°C
UV spectrophotometry>=92 %
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen), Radical initiators
RUO – Research Use Only