Dibenzyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Dibenzyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dibenzyl azodicarboxylate, 94%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 270270250
Price (USD)/ Each
181.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
213.00 
Save 31.35 (15%)
-
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
181.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
213.00 
Save 31.35 (15%)
Add to cart
Dibenzyl azodicarboxylate, 94%, Thermo Scientific Chemicals
Catalog number270270250
Price (USD)/ Each
181.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
213.00 
Save 31.35 (15%)
-
Add to cart
Chemical Identifiers
IUPAC NameN-{[(benzyloxy)carbonyl]imino}(benzyloxy)formamide
Molecular FormulaC16H14N2O4
InChI KeyIRJKSAIGIYODAN-UHFFFAOYSA-N
SMILESO=C(OCC1=CC=CC=C1)N=NC(=O)OCC1=CC=CC=C1
Molecular Weight (g/mol)298.30
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to orange
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point44°C to 52°C
UV spectrophotometry>=92 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X Bond Formation (Halogen), Radical initiators
RUO – Research Use Only