Formic acid, 99%, for analysis
Formic acid, 99%, for analysis
Formic acid, 99%, for analysis
Formic acid, 99%, for analysis
Thermo Scientific Chemicals

Formic acid, 99%, for analysis

Formic acid, 99%, CH2O2, CAS Number-64-18-6, acide formique, aminic acid, bilorin, collo-bueglatt, formira, formisoton, formylic acid, hydrogen carboxylic acid, methanoic acid, myrmicyl, 2.5L, Vapor Pressure: 44mbar at 20 deg.C, 02, 8, 101.0 deg.C, 0.1ppm max., CHEBI:30751, 5ppm max., Undesignated | CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Quantity:
25 mL
1 L
10 L
Packaging:
View all
Plastic Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 270480250
Price (JPY)
-
Quantity:
25 mL
Packaging:
Plastic Bottle
Request bulk or custom format
Chemical Identifiers
CAS64-18-6
IUPAC Nameformic acid
Molecular FormulaCH2O2
InChI KeyBDAGIHXWWSANSR-UHFFFAOYSA-N
SMILESOC=O
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Dilution testpasses test
Chloride (Cl)=<5 ppm
Sulfite (SO3)=<20 ppm
Cadmium (Cd)=<0.1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only